Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191023.1 | NP_001177952.1 | 1 - 2 | 298 | 68 |
NM_001191025.1 | NP_001177954.1 | 1 - 2 | 298 | 68 | |
NM_001191026.1 | NP_001177955.1 | 1 - 2 | 298 | 68 | |
NM_001191027.1 | NP_001177956.1 | 1 - 2 | 298 | 68 | |
NM_010063.4 | NP_034193.2 | 1 - 2 | 298 | 68 | |
GenBank mRNA | AK158589.1 | - | 1 - 2 | 299 | 68 |
BY238976.1 | - | 1 - 2 | 147 | 68 |