Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191023.1 | NP_001177952.1 | 5 - 6 | 784 | 66 |
NM_001191025.1 | NP_001177954.1 | 5 - 6 | 733 | 66 | |
NM_001191026.1 | NP_001177955.1 | 6 - 7 | 760 | 66 | |
NM_001191027.1 | NP_001177956.1 | 5 - 6 | 700 | 66 | |
NM_010063.4 | NP_034193.2 | 6 - 7 | 793 | 66 | |
GenBank mRNA | AF063229.1 | - | 6 - 7 | 627 | 66 |
AF063230.1 | - | 5 - 6 | 567 | 66 | |
AK158589.1 | - | 6 - 7 | 761 | 66 | |
BC051992.1 | - | 6 - 7 | 521 | 66 | |
GU992206.1 | - | 5 - 6 | 525 | 66 | |
GU992207.1 | - | 5 - 6 | 441 | 66 |