Assay Design



Gene Transcripts

Gene Symbol
Gpld1
Entrez Gene ID
14756
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
6330541J12Rik, AW546131
Chromosome Location
Chr.13: 24943152 - 24991936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008156.2 NP_032182.2 22 - 23 2518 78
GenBank mRNA AF050666.1 - 22 - 23 2457 78
AK004957.2 - 21 - 22 2222 78
AY081194.1 - 21 - 22 2245 78
AY333960.1 - 21 - 22 2202 78
BC019146.1 - 21 - 22 2231 78

View Full Product Details