Assay Design



Gene Transcripts

Gene Symbol
Gdpd5
Entrez Gene ID
233552
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
BC024955, Gde2
Chromosome Location
Chr.7: 99381415 - 99460984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201352.2 NP_958740.2 16 - 17 2449 115
GenBank mRNA AK141189.1 - 16 - 17 2482 115
AK154771.1 - 16 - 17 2586 115
BC024955.1 - 15 - 16 1735 115
BC026428.1 - 12 - 13 1382 115
BC082585.1 - 16 - 17 2449 115

View Full Product Details