Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011035.2 | NP_035165.2 | 6 - 7 | 824 | 67 |
GenBank mRNA | AF082077.1 | - | 6 - 7 | 789 | 67 |
AK077497.1 | - | 6 - 7 | 790 | 67 | |
BC079838.1 | - | 6 - 7 | 810 | 67 | |
BC145698.1 | - | 7 - 8 | 748 | 67 |