Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011035.2 | NP_035165.2 | 9 - 10 | 1113 | 100 |
GenBank mRNA | AF082077.1 | - | 9 - 10 | 1078 | 100 |
AK077497.1 | - | 9 - 10 | 1079 | 100 | |
BC079838.1 | - | 9 - 10 | 1098 | 100 | |
BC145698.1 | - | 10 - 11 | 1037 | 100 |