Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285870.1 | NP_001272799.1 | 6 - 7 | 1656 | 100 |
NM_001285871.1 | NP_001272800.1 | 6 - 7 | 1841 | 100 | |
NM_001285873.1 | NP_001272802.1 | 6 - 7 | 1656 | 100 | |
NM_018880.3 | NP_061368.1 | 6 - 7 | 1630 | 100 | |
GenBank mRNA | AB030912.1 | - | 6 - 7 | 1657 | 100 |
AF220019.1 | - | 6 - 7 | 1549 | 100 | |
AK019165.1 | - | 6 - 7 | 1591 | 100 | |
AK144674.1 | - | 6 - 7 | 1559 | 100 | |
AK170029.1 | - | 6 - 7 | 1594 | 100 | |
BC003347.1 | - | 6 - 7 | 1670 | 100 | |
BC034263.1 | - | 6 - 7 | 1595 | 100 |