Assay Design



Gene Transcripts

Gene Symbol
Asns
Entrez Gene ID
27053
Gene Name
asparagine synthetase
Gene Aliases
-
Chromosome Location
Chr.6: 7675165 - 7693182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012055.3 NP_036185.1 4 - 5 774 65
GenBank mRNA AK076207.1 - 4 - 5 792 65
AK160459.1 - 4 - 5 789 65
AK167524.1 - 4 - 5 754 65
AK167933.1 - 4 - 5 773 65
BC005552.1 - 4 - 5 772 65
BC050123.1 - - 3102 65
U38940.1 - 4 - 5 775 65

View Full Product Details