Assay Design



Gene Transcripts

Gene Symbol
Umod
Entrez Gene ID
22242
Gene Name
uromodulin
Gene Aliases
THP, Urehd1, urehr4
Chromosome Location
Chr.7: 119462708 - 119479262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278605.1 NP_001265534.1 5 - 6 1329 80
NM_009470.5 NP_033496.1 5 - 6 1385 80
GenBank mRNA AK085460.1 - 5 - 6 1379 80
AK144065.1 - 5 - 6 1301 80
AK164688.1 - 5 - 6 1301 80
AK165507.1 - 5 - 6 1245 80
BC012973.1 - 4 - 5 1223 80
L33406.1 - 5 - 6 1315 80

View Full Product Details