Assay Design



Gene Transcripts

Gene Symbol
Hirip3
Entrez Gene ID
233876
Gene Name
HIRA interacting protein 3
Gene Aliases
B130036O03, C86302
Chromosome Location
Chr.7: 126861972 - 126865377 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172746.3 NP_766334.1 1 - 2 545 91
GenBank mRNA AK045122.1 - 1 - 2 80 91
AK135526.1 - 1 - 2 88 91
BC055687.1 - 1 - 2 545 91

View Full Product Details