Assay Design



Gene Transcripts

Gene Symbol
Ano1
Entrez Gene ID
101772
Gene Name
anoctamin 1, calcium activated chloride channel
Gene Aliases
Tmem16a
Chromosome Location
Chr.7: 144588549 - 144752039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242349.1 NP_001229278.1 5 - 6 966 88
NM_178642.5 NP_848757.4 5 - 6 963 88
GenBank mRNA AK052589.1 - 5 - 6 915 88
BC062959.1 - 5 - 6 852 88

View Full Product Details