Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028841.3 | NP_083117.2 | 1 - 2 | 279 | 77 |
GenBank mRNA | AK008761.1 | - | 1 - 2 | 224 | 77 |
AK166272.1 | - | 1 - 2 | 282 | 77 | |
AK171691.1 | - | 1 - 2 | 228 | 77 | |
BC010346.1 | - | 1 - 2 | 224 | 77 | |
BY734818.1 | - | 1 - 2 | 279 | 77 |