Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252588.1 | NP_001239517.1 | 6 - 7 | 680 | 72 |
NM_053082.3 | NP_444312.1 | 6 - 7 | 843 | 72 | |
GenBank mRNA | AK002709.1 | - | 6 - 7 | 603 | 72 |
AK156959.1 | - | 6 - 7 | 799 | 72 | |
AK172481.1 | - | 6 - 7 | 661 | 72 | |
AK217510.1 | - | 2 - 3 | 171 | 72 | |
AK218064.1 | - | 2 - 3 | 173 | 72 | |
BC003482.1 | - | 6 - 7 | 623 | 72 | |
BC083070.1 | - | 6 - 7 | 839 | 72 | |
CT010285.1 | - | 5 - 6 | 564 | 72 |