Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019553.2 | NP_062426.2 | 10 - 11 | 1971 | 102 |
GenBank mRNA | AF159131.3 | - | 10 - 11 | 1932 | 102 |
AK010281.1 | - | 10 - 11 | 1851 | 102 | |
AK145578.1 | - | 10 - 11 | 1955 | 102 | |
AK160095.1 | - | 10 - 11 | 1945 | 102 | |
BC030895.1 | - | 9 - 10 | 1403 | 102 | |
BC043655.1 | - | 10 - 11 | 1943 | 102 | |
BC059237.1 | - | 10 - 11 | 1931 | 102 |