Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163276.1 | NP_001156748.1 | 4 - 5 | 566 | 70 |
NM_198107.2 | NP_932775.2 | 4 - 5 | 569 | 70 | |
GenBank mRNA | AK042340.1 | - | 4 - 5 | 531 | 70 |
AK138125.1 | - | 4 - 5 | 553 | 70 | |
AK149345.1 | - | 4 - 5 | 550 | 70 | |
BC057056.1 | - | 4 - 5 | 523 | 70 |