Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163276.1 | NP_001156748.1 | 6 - 7 | 1099 | 70 |
NM_198107.2 | NP_932775.2 | 6 - 7 | 1102 | 70 | |
GenBank mRNA | AK042340.1 | - | 6 - 7 | 1064 | 70 |
AK149345.1 | - | 6 - 7 | 1083 | 70 | |
BC057056.1 | - | 6 - 7 | 1056 | 70 |