Assay Design



Gene Transcripts

Gene Symbol
Boc
Entrez Gene ID
117606
Gene Name
biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein
Gene Aliases
4732455C11, mFLJ00376
Chromosome Location
Chr.16: 44485045 - 44558870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172506.2 NP_766094.1 12 - 13 2716 109
GenBank mRNA AF388037.1 - 11 - 12 2169 109
AK028770.1 - 12 - 13 2737 109
AK028889.1 - 12 - 13 2717 109
AK131182.1 - 7 - 8 4936 109
BC026443.1 - 1 - 2 117 109
BC056138.1 - 3 - 4 619 109
BC062892.1 - 12 - 13 2696 109
BC078631.1 - 12 - 13 2660 109

View Full Product Details