Assay Design



Gene Transcripts

Gene Symbol
Rfx7
Entrez Gene ID
319758
Gene Name
regulatory factor X, 7
Gene Aliases
2510005N23Rik, 9930116O05Rik, D130086K05Rik, Rfxdc2
Chromosome Location
Chr.9: 72532167 - 72622949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033536.1 NP_001028708.1 5 - 6 1105 82
GenBank mRNA AK090370.1 - 5 - 6 1106 82
AK170249.1 - 6 - 7 577 82

View Full Product Details