Assay Design



Gene Transcripts

Gene Symbol
Slc12a8
Entrez Gene ID
171286
Gene Name
solute carrier family 12 (potassium/chloride transporters), member 8
Gene Aliases
CCC9, E330020C02Rik
Chromosome Location
Chr.16: 33517333 - 33664135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083902.1 NP_001077371.1 1 - 2 81 115
GenBank mRNA BC030926.1 - 1 - 2 81 115

View Full Product Details