Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010925.2 | NP_035055.2 | 10 - 11 | 1250 | 72 |
GenBank mRNA | AF312394.1 | - | 9 - 10 | 1013 | 72 |
AK077540.1 | - | 10 - 11 | 1175 | 72 | |
BC003913.1 | - | 3 - 4 | 346 | 72 | |
BC022910.1 | - | 9 - 10 | 1116 | 72 | |
BC065993.1 | - | 10 - 11 | 1212 | 72 | |
BC065994.1 | - | 10 - 11 | 1170 | 72 | |
U79773.1 | - | 11 - 12 | 1049 | 72 | |
U79774.1 | - | 10 - 11 | 1202 | 72 |