Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173413.3 | NP_775589.1 | 4 - 5 | 391 | 118 |
GenBank mRNA | AB232601.1 | - | 4 - 5 | 319 | 118 |
AK051218.1 | - | 4 - 5 | 341 | 118 | |
AK084650.1 | - | 4 - 5 | 341 | 118 | |
AK155621.1 | - | 4 - 5 | 341 | 118 | |
BC059208.1 | - | 4 - 5 | 373 | 118 | |
BC157974.1 | - | 4 - 5 | 339 | 118 |