Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007585.3 | NP_031611.1 | 12 - 13 | 1036 | 69 |
GenBank mRNA | AK012563.1 | - | 13 - 14 | 1038 | 69 |
AK088645.1 | - | 12 - 13 | 1025 | 69 | |
AK146096.1 | - | 12 - 13 | 1024 | 69 | |
AK150586.1 | - | 12 - 13 | 1024 | 69 | |
AK152561.1 | - | 12 - 13 | 1024 | 69 | |
AK160746.1 | - | 12 - 13 | 1024 | 69 | |
AK164507.1 | - | 12 - 13 | 1029 | 69 | |
AK165888.1 | - | 12 - 13 | 1024 | 69 | |
AK167894.1 | - | 12 - 13 | 1023 | 69 | |
AK168083.1 | - | 12 - 13 | 1024 | 69 | |
BC003327.1 | - | 12 - 13 | 1036 | 69 | |
BC004659.1 | - | 8 - 9 | 1932 | 69 | |
BC005763.1 | - | 12 - 13 | 1010 | 69 | |
D10024.1 | - | 11 - 12 | 969 | 69 | |
M14044.1 | - | 11 - 12 | 969 | 69 |