Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019686.5 | NP_062660.1 | 4 - 5 | 405 | 86 |
GenBank mRNA | AB016080.1 | - | 4 - 5 | 405 | 86 |
AK005347.1 | - | 4 - 5 | 509 | 86 | |
AK013240.1 | - | - | 514 | 86 | |
BC005739.1 | - | 4 - 5 | 447 | 86 |