Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110305.1 | NP_001103775.1 | 5 - 6 | 2294 | 69 |
NM_001110306.1 | NP_001103776.1 | 5 - 6 | 2179 | 69 | |
NM_001110307.1 | NP_001103777.1 | 5 - 6 | 2011 | 69 | |
NM_016679.4 | NP_057888.1 | 4 - 5 | 3330 | 69 | |
GenBank mRNA | AB020063.1 | - | 5 - 6 | 1783 | 69 |
AK004738.1 | - | 5 - 6 | 2011 | 69 | |
AK039549.1 | - | 5 - 6 | 1950 | 69 | |
AK046178.1 | - | 5 - 6 | 1895 | 69 | |
AK076234.1 | - | 4 - 5 | 2816 | 69 | |
AK129061.1 | - | 4 - 5 | 1765 | 69 | |
AK136319.1 | - | 3 - 4 | 1015 | 69 | |
AK150437.1 | - | 5 - 6 | 1915 | 69 | |
AK150485.1 | - | 5 - 6 | 1782 | 69 | |
AK152142.1 | - | 5 - 6 | 1915 | 69 | |
AK159858.1 | - | 5 - 6 | 1780 | 69 | |
BC055732.1 | - | 4 - 5 | 1768 | 69 |