Assay Design



Gene Transcripts

Gene Symbol
Tex9
Entrez Gene ID
21778
Gene Name
testis expressed gene 9
Gene Aliases
Gm19407, tsec-1
Chromosome Location
Chr.9: 72458055 - 72510944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009359.4 NP_033385.2 3 - 4 183 56
GenBank mRNA AB000619.1 - 4 - 5 431 56
AK008505.1 - 3 - 4 181 56
AK012189.1 - 3 - 4 182 56
AK018568.1 - - 181 56
AK032490.1 - 2 - 3 290 56
AK141174.1 - 2 - 3 293 56
BC141033.1 - 3 - 4 325 56
BC145184.1 - 3 - 4 325 56
BC145185.1 - 3 - 4 325 56
BC145186.1 - 3 - 4 325 56

View Full Product Details