Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285493.1 | NP_001272422.1 | 1 - 2 | 79 | 58 |
NM_001285494.1 | NP_001272423.1 | 1 - 2 | 79 | 58 | |
NM_023120.5 | NP_075609.2 | 1 - 2 | 79 | 58 | |
NR_104347.1 | 1 - 2 | 79 | 58 | ||
GenBank mRNA | AK158210.1 | - | 1 - 2 | 66 | 58 |
AK173227.1 | - | 1 - 2 | 62 | 58 | |
BC024635.1 | - | 1 - 2 | 48 | 58 | |
BC029862.1 | - | 1 - 2 | 49 | 58 |