Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290497.1 | NP_001277426.1 | 1 - 2 | 138 | 88 |
NM_001290498.1 | NP_001277427.1 | 1 - 2 | 138 | 88 | |
NM_177102.4 | NP_796076.1 | 1 - 2 | 138 | 88 | |
GenBank mRNA | AK043858.1 | - | 1 - 2 | 91 | 88 |
AK079312.1 | - | 1 - 2 | 139 | 88 | |
AK086094.1 | - | 1 - 2 | 90 | 88 | |
BC058996.1 | - | 1 - 2 | 87 | 88 | |
BQ555196.1 | - | 1 - 2 | 66 | 88 |