Assay Design
Gene Transcripts
- Gene Symbol
- Rtkn2
- Entrez Gene ID
-
170799
- Gene Name
- rhotekin 2
- Gene Aliases
-
B130039D23Rik, Mbf, Plekhk1
- Chromosome Location
-
Chr.10: 67979566 - 68043864 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC116341.2 |
- |
- |
1334 |
90 |
BC116342.2 |
- |
- |
1334 |
90 |
BC158093.1 |
- |
10 - 11 |
1337 |
90 |
View Full Product Details