Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008591.2 | NP_032617.2 | 16 - 17 | 3606 | 74 |
GenBank mRNA | BC117969.1 | - | 14 - 15 | 3250 | 74 |
BC117970.1 | - | 14 - 15 | 3246 | 74 | |
BU514228.1 | - | 2 - 3 | 263 | 74 | |
Y00671.1 | - | 14 - 15 | 3251 | 74 |