Assay Design



Gene Transcripts

Gene Symbol
Met
Entrez Gene ID
17295
Gene Name
met proto-oncogene
Gene Aliases
AI838057, HGF, HGFR, Par4, c-Met
Chromosome Location
Chr.6: 17463349 - 17573980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008591.2 NP_032617.2 9 - 10 2460 123
GenBank mRNA AK148258.1 - 9 - 10 2313 123
BC117969.1 - 7 - 8 2104 123
BC117970.1 - 7 - 8 2100 123
CK625647.1 - 1 - 2 137 123
Y00671.1 - 7 - 8 2105 123

View Full Product Details