Assay Design



Gene Transcripts

Gene Symbol
Chchd3
Entrez Gene ID
66075
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
0610041L09Rik, 1700039J09Rik, AW558177
Chromosome Location
Chr.6: 32792217 - 33060276 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025336.1 NP_079612.1 3 - 4 323 81
GenBank mRNA AK002263.1 - 3 - 4 388 81
AK002889.1 - 3 - 4 395 81
AK003519.1 - 3 - 4 323 81
AK006643.1 - 3 - 4 389 81
AK145337.1 - 3 - 4 372 81
AK193067.1 - 2 - 3 149 81
AK217481.1 - 2 - 3 149 81
BC016528.1 - 2 - 3 145 81
BC021941.1 - 3 - 4 375 81
BC032987.1 - 3 - 4 338 81

View Full Product Details