Assay Design
Gene Transcripts
- Gene Symbol
- March11
- Entrez Gene ID
-
211147
- Gene Name
- membrane-associated ring finger (C3HC4) 11
- Gene Aliases
-
9630025C22
- Chromosome Location
-
Chr.15: 26308904 - 26409594 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177597.5 |
NP_808265.2 |
3 - 4 |
1063 |
98 |
View Full Product Details