Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007462.3 | NP_031488.2 | 1 - 2 | 288 | 150 |
GenBank mRNA | AK042021.1 | - | 1 - 2 | 289 | 150 |
AK045053.1 | - | 1 - 2 | 259 | 150 | |
AK046280.1 | - | 1 - 2 | 279 | 150 | |
AK080907.1 | - | 1 - 2 | 288 | 150 |