Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181414.5 | NP_852079.2 | 12 - 13 | 1468 | 98 |
GenBank mRNA | AK030578.1 | - | 12 - 13 | 1498 | 98 |
AK044997.1 | - | 12 - 13 | 1468 | 98 | |
BC027029.1 | - | 12 - 13 | 1437 | 98 | |
BC057678.1 | - | 12 - 13 | 1471 | 98 |