Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027366.1 | NP_081642.1 | 2 - 3 | 390 | 59 |
GenBank mRNA | AJ315550.1 | - | 2 - 3 | 177 | 59 |
AK009303.1 | - | 2 - 3 | 247 | 59 | |
BC132420.1 | - | 2 - 3 | 238 | 59 | |
BC138777.1 | - | 2 - 3 | 238 | 59 |