Assay Design



Gene Transcripts

Gene Symbol
Ly6g6d
Entrez Gene ID
114654
Gene Name
lymphocyte antigen 6 complex, locus G6D
Gene Aliases
A930024F17Rik, G6d, G6f, MEGT1, NG25, NG32
Chromosome Location
Chr.17: 35071342 - 35080567 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033478.2 NP_258439.1 1 - 2 53 62
GenBank mRNA AJ315548.1 - 1 - 2 53 62
BC131988.1 - 1 - 2 89 62
BC131990.1 - 1 - 2 89 62

View Full Product Details