Assay Design



Gene Transcripts

Gene Symbol
Ddx39b
Entrez Gene ID
53817
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B
Gene Aliases
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh
Chromosome Location
Chr.17: 35241746 - 35253707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019693.3 NP_062667.1 1 - 2 103 101
GenBank mRNA AK051034.1 - 1 - 2 36 101
AK171507.1 - 1 - 2 45 101
CN716634.1 - 1 - 2 98 101

View Full Product Details