Assay Design



Gene Transcripts

Gene Symbol
Spon1
Entrez Gene ID
233744
Gene Name
spondin 1, (f-spondin) extracellular matrix protein
Gene Aliases
AI666765, AW455831, BC020531, D330035F22Rik, FSP
Chromosome Location
Chr.7: 113765998 - 114043375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145584.2 NP_663559.1 7 - 8 1225 94
GenBank mRNA AK146506.1 - 7 - 8 1050 94
AK173021.1 - 7 - 8 892 94
BC020531.1 - 7 - 8 1095 94

View Full Product Details