Assay Design
Gene Transcripts
- Gene Symbol
- Sft2d3
- Entrez Gene ID
-
67158
- Gene Name
- SFT2 domain containing 3
- Gene Aliases
-
2610206C24Rik, AI847965, AW046173
- Chromosome Location
-
Chr.18: 31909094 - 31911903 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026006.1 |
NP_080282.1 |
1 - 1 |
2503 |
93 |
GenBank mRNA |
AK011898.1 |
- |
1 - 1 |
1878 |
93 |
View Full Product Details