Assay Design



Gene Transcripts

Gene Symbol
Hspa1a
Entrez Gene ID
193740
Gene Name
heat shock protein 1A
Gene Aliases
Hsp70-3, Hsp70.3, Hsp72, hsp68, hsp70A1
Chromosome Location
Chr.17: 34969359 - 34972156 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010479.2 NP_034609.2 1 - 1 2357 121
GenBank mRNA AK132331.1 - 1 - 1 2357 121
BC054782.1 - 1 - 1 2325 121

View Full Product Details