Assay Design



Gene Transcripts

Gene Symbol
Mmp19
Entrez Gene ID
58223
Gene Name
matrix metallopeptidase 19
Gene Aliases
-
Chromosome Location
Chr.10: 128790910 - 128800824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164197.1 NP_001157669.1 3 - 4 277 73
NM_021412.2 NP_067387.1 3 - 4 412 73
GenBank mRNA AF153199.1 - 3 - 4 364 73
AF155221.1 - 3 - 4 392 73
AF162446.1 - 3 - 4 392 73
AK079924.1 - 3 - 4 278 73
AK080125.1 - 3 - 4 272 73
AK086808.1 - 2 - 3 152 73
AK136517.1 - 3 - 4 266 73
AK154863.1 - 3 - 4 405 73
BC112966.2 - 3 - 4 384 73

View Full Product Details