Assay Design



Gene Transcripts

Gene Symbol
Erbb3
Entrez Gene ID
13867
Gene Name
v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Aliases
C76256, Erbb-3, Erbb3r, Her3
Chromosome Location
Chr.10: 128569368 - 128589501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010153.1 NP_034283.1 1 - 2 86 60
GenBank mRNA AY686636.1 - 1 - 2 86 60
BC106091.1 - - 237 60

View Full Product Details