Assay Design



Gene Transcripts

Gene Symbol
Il23a
Entrez Gene ID
83430
Gene Name
interleukin 23, alpha subunit p19
Gene Aliases
IL-23, p19
Chromosome Location
Chr.10: 128296140 - 128298084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031252.2 NP_112542.1 1 - 2 269 109
GenBank mRNA AF301619.1 - 1 - 2 269 109
BC019953.1 - 1 - 2 253 109

View Full Product Details