Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286015.1 | NP_001272944.1 | 27 - 28 | 740 | 95 |
NM_001286016.1 | NP_001272945.1 | 28 - 29 | 764 | 95 | |
NM_007784.3 | NP_031810.1 | 28 - 29 | 764 | 95 | |
GenBank mRNA | AK085690.1 | - | 28 - 29 | 768 | 95 |
BC002101.1 | - | 28 - 29 | 733 | 95 | |
BC003859.1 | - | 28 - 29 | 748 | 95 | |
BC006024.1 | - | 27 - 28 | 724 | 95 | |
BC007192.1 | - | 28 - 29 | 748 | 95 | |
BC008278.1 | - | 28 - 29 | 748 | 95 | |
BC020321.1 | - | 9 - 10 | 218 | 95 | |
BC040246.1 | - | 28 - 29 | 760 | 95 | |
M36780.1 | - | 27 - 28 | 703 | 95 |