Assay Design
Gene Transcripts
- Gene Symbol
- Tcaf3
- Entrez Gene ID
-
403088
- Gene Name
- TRPM8 channel-associated factor 3
- Gene Aliases
-
Eapa2, Fam115e
- Chromosome Location
-
Chr.6: 42587213 - 42597372 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203396.1 |
NP_981933.1 |
1 - 2 |
716 |
130 |
GenBank mRNA |
AY528666.1 |
- |
1 - 2 |
716 |
130 |
View Full Product Details