Assay Design



Gene Transcripts

Gene Symbol
Gpank1
Entrez Gene ID
81845
Gene Name
G patch domain and ankyrin repeats 1
Gene Aliases
Bat-4, Bat4, D17H6S54E, G5
Chromosome Location
Chr.17: 35121473 - 35124815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128597.1 NP_001122069.1 1 - 2 302 110
GenBank mRNA BC057902.1 - 1 - 2 261 110
CJ069410.1 - 1 - 2 305 110

View Full Product Details