Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146041.2 | NP_666153.1 | 3 - 4 | 301 | 127 |
GenBank mRNA | AK086076.1 | - | 3 - 4 | 343 | 127 |
BC031788.1 | - | 3 - 4 | 301 | 127 | |
BC093502.1 | - | 3 - 4 | 325 | 127 |