Assay Design



Gene Transcripts

Gene Symbol
Nrf1
Entrez Gene ID
18181
Gene Name
nuclear respiratory factor 1
Gene Aliases
C87038, D6Ertd415e
Chromosome Location
Chr.6: 30047988 - 30153458 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164227.1 NP_001157699.1 11 - 12 1640 69
GenBank mRNA AK037697.1 - 11 - 12 1598 69
BC005410.1 - 10 - 11 1458 69

View Full Product Details