Assay Design



Gene Transcripts

Gene Symbol
Slc17a3
Entrez Gene ID
105355
Gene Name
solute carrier family 17 (sodium phosphate), member 3
Gene Aliases
AW261723, Npt4
Chromosome Location
Chr.13: 23836918 - 23860718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164743.1 NP_001158215.1 9 - 10 1415 157
NM_134069.3 NP_598830.2 10 - 11 1539 157
GenBank mRNA AK128896.1 - 10 - 11 1523 157
AK143896.1 - 10 - 11 1511 157
BC018252.1 - 10 - 11 1468 157
BC024459.1 - 10 - 11 1495 157
BC034179.1 - 10 - 11 1359 157
BC034183.1 - 10 - 11 1344 157

View Full Product Details