Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028292.2 | NP_082568.1 | 3 - 4 | 363 | 70 |
GenBank mRNA | AK012256.1 | - | 3 - 4 | 435 | 70 |
AK028261.1 | - | 3 - 4 | 398 | 70 | |
AK076992.1 | - | 3 - 4 | 774 | 70 | |
AK146051.1 | - | 3 - 4 | 345 | 70 | |
AK146268.1 | - | 3 - 4 | 375 | 70 | |
AK154876.1 | - | 3 - 4 | 436 | 70 | |
BC014867.1 | - | 3 - 4 | 363 | 70 |